Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hpca
Entrez Gene ID
100750673
Gene Name
hippocalcin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613801: 1307710 - 1313710 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500628.1 XP_003500676.1 2 - 3 486 72

View Full Product Details