Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lum
Entrez Gene ID
100755861
Gene Name
lumican
Gene Aliases
I79_010810
Chromosome Location
Chr.Un|NW_003614008: 373484 - 381166 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503874.2 XP_003503922.1 2 - 3 1302 68

View Full Product Details