Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serf2
Entrez Gene ID
100754083
Gene Name
small EDRK-rich factor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613677: 853915 - 855507 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498032.2 XP_003498080.1 2 - 3 198 93
GenBank mRNA JI863445.1 - 2 - 3 176 93

View Full Product Details