Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qpct
Entrez Gene ID
100755692
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614440: 533103 - 570163 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507855.1 XP_003507903.1 2 - 3 269 93

View Full Product Details