Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fem1b
Entrez Gene ID
100761263
Gene Name
fem-1 homolog b (C. elegans)
Gene Aliases
I79_006761
Chromosome Location
Chr.Un|NW_003613796: 1411207 - 1450636 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641253.1 XP_007639443.1 2 - 3 571 69

View Full Product Details