Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxcl11
Entrez Gene ID
100771051
Gene Name
chemokine (C-X-C motif) ligand 11
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615570: 105387 - 107338 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653171.1 XP_007651361.1 3 - 4 260 79

View Full Product Details