Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nrn1
Entrez Gene ID
100761420
Gene Name
neuritin 1
Gene Aliases
I79_003079, Neuritin
Chromosome Location
Chr.Un|NW_003613650: 2519061 - 2527868 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497478.2 XP_003497526.1 1 - 2 228 107

View Full Product Details