Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lemd1
Entrez Gene ID
100760186
Gene Name
LEM domain containing 1
Gene Aliases
I79_007589
Chromosome Location
Chr.Un|NW_003613838: 770735 - 803091 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501222.1 XP_003501270.1 4 - 5 344 70

View Full Product Details