Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gfi1
Entrez Gene ID
100769674
Gene Name
growth factor independent 1 transcription repressor
Gene Aliases
I79_008906
Chromosome Location
Chr.Un|NW_003613892: 661591 - 670426 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642991.1 XP_007641181.1 7 - 8 996 82

View Full Product Details