Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx4
Entrez Gene ID
100750849
Gene Name
LIM homeobox 4
Gene Aliases
I79_006317
Chromosome Location
Chr.Un|NW_003613774: 1549474 - 1589302 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500115.2 XP_003500163.1 5 - 6 777 77

View Full Product Details