Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrnb2
Entrez Gene ID
100761068
Gene Name
cholinergic receptor, nicotinic, beta 2 (neuronal)
Gene Aliases
I79_008026
Chromosome Location
Chr.Un|NW_003613854: 929737 - 939057 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501604.2 XP_003501652.1 1 - 2 110 62

View Full Product Details