Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a1
Entrez Gene ID
100766982
Gene Name
solute carrier family 39 (zinc transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613854: 1420956 - 1425030 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501629.2 XP_003501677.1 1 - 2 458 103
XM_007642290.1 XP_007640480.1 2 - 3 367 103
XM_007642291.1 XP_007640481.1 2 - 3 658 103

View Full Product Details