Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nudt2
Entrez Gene ID
100769146
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615063: 92692 - 102899 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511230.2 XP_003511278.1 3 - 4 518 77
XM_007651763.1 XP_007649953.1 4 - 5 628 77
XM_007651764.1 XP_007649954.1 4 - 5 610 77
XM_007651765.1 XP_007649955.1 4 - 5 623 77
GenBank mRNA JI874705.1 - 1 - 2 168 77

View Full Product Details