Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Egln1
Entrez Gene ID
100772704
Gene Name
egl-9 family hypoxia-inducible factor 1
Gene Aliases
I79_002290
Chromosome Location
Chr.Un|NW_003613636: 2145857 - 2183052 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653534.1 XP_007651724.1 2 - 3 440 101

View Full Product Details