Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nsrp1
Entrez Gene ID
100754999
Gene Name
nuclear speckle splicing regulatory protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614210: 495641 - 521523 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506083.2 XP_003506131.1 3 - 4 867 67
XM_007646553.1 XP_007644743.1 3 - 4 225 67
GenBank mRNA JP059673.1 - 1 - 2 62 67

View Full Product Details