Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp21
Entrez Gene ID
100756697
Gene Name
matrix metallopeptidase 21
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616182: 56697 - 64188 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513871.2 XP_003513919.1 2 - 3 1194 81
XM_007654102.1 XP_007652292.1 2 - 3 1194 81

View Full Product Details