Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl20
Entrez Gene ID
100756991
Gene Name
chemokine (C-C motif) ligand 20
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617185: 27843 - 30996 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514973.2 XP_003515021.1 1 - 2 130 84

View Full Product Details