Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcd7
Entrez Gene ID
100753946
Gene Name
programmed cell death 7
Gene Aliases
I79_014761
Chromosome Location
Chr.Un|NW_003614337: 553912 - 562616 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647718.1 XP_007645908.1 4 - 5 644 55
GenBank mRNA JP043458.1 - 4 - 5 537 55

View Full Product Details