Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shc4
Entrez Gene ID
100763505
Gene Name
SHC (Src homology 2 domain containing) family, member 4
Gene Aliases
I79_013551
Chromosome Location
Chr.Un|NW_003614219: 166972 - 267130 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506251.2 XP_003506299.1 11 - 12 2324 64

View Full Product Details