Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abo
Entrez Gene ID
100772592
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Gene Aliases
I79_020335
Chromosome Location
Chr.Un|NW_003615220: 199278 - 209434 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652261.1 XP_007650451.1 4 - 5 476 78

View Full Product Details