Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl9
Entrez Gene ID
100759276
Gene Name
ADP-ribosylation factor-like 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615992: 92887 - 104068 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513529.2 XP_003513577.1 3 - 4 1256 119
XM_007653816.1 XP_007652006.1 3 - 4 426 119
GenBank mRNA JI877116.1 - 3 - 4 1295 119
JP054182.1 - 3 - 4 1295 119

View Full Product Details