Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Satl1
Entrez Gene ID
100756097
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
I79_021879
Chromosome Location
Chr.Un|NW_003615684: 126895 - 137507 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653360.1 XP_007651550.1 7 - 8 2092 80

View Full Product Details