Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Npy1r
Entrez Gene ID
100757157
Gene Name
neuropeptide Y receptor Y1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614963: 127390 - 128646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510777.1 XP_003510825.1 1 - 2 693 77

View Full Product Details