Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd39
Entrez Gene ID
100769174
Gene Name
ankyrin repeat domain 39
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613687: 956845 - 967297 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498294.2 XP_003498342.1 1 - 2 141 64

View Full Product Details