Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spx
Entrez Gene ID
100773420
Gene Name
spexin hormone
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613944: 573516 - 579548 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503173.1 XP_003503221.1 4 - 5 283 100

View Full Product Details