Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fpr2
Entrez Gene ID
100758340
Gene Name
formyl peptide receptor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616796: 19433 - 20485 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514633.1 XP_003514681.1 1 - 1 1012 68

View Full Product Details