Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ly6g5b
Entrez Gene ID
100771711
Gene Name
lymphocyte antigen 6 complex, locus G5B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614548: 493967 - 495004 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649116.1 XP_007647306.1 2 - 3 142 59

View Full Product Details