Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Coq10a
Entrez Gene ID
100750875
Gene Name
coenzyme Q10 homolog A (S. cerevisiae)
Gene Aliases
I79_013491
Chromosome Location
Chr.Un|NW_003614217: 428869 - 436234 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646682.1 XP_007644872.1 1 - 2 470 77

View Full Product Details