Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc17
Entrez Gene ID
100761042
Gene Name
coiled-coil domain containing 17
Gene Aliases
I79_002951
Chromosome Location
Chr.Un|NW_003613647: 2162660 - 2168710 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007611633.1 XP_007609823.1 11 - 12 3153 67

View Full Product Details