Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aqp1
Entrez Gene ID
100773231
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614013: 996417 - 1008942 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503923.2 XP_003503971.1 3 - 4 705 98
XM_007644456.1 XP_007642646.1 3 - 4 431 98
XM_007644457.1 XP_007642647.1 3 - 4 492 98

View Full Product Details