Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1qtnf4
Entrez Gene ID
100764222
Gene Name
C1q and tumor necrosis factor related protein 4
Gene Aliases
I79_002895
Chromosome Location
Chr.Un|NW_003613646: 2799307 - 2800160 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007610659.1 XP_007608849.1 1 - 2 153 57

View Full Product Details