Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zmym5
Entrez Gene ID
100751650
Gene Name
zinc finger, MYM-type 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613875: 63777 - 78304 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642655.1 XP_007640845.1 4 - 5 956 173

View Full Product Details