Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl7
Entrez Gene ID
100755012
Gene Name
F-box and leucine-rich repeat protein 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614545: 60127 - 410519 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649078.1 XP_007647268.1 3 - 4 753 63

View Full Product Details