Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lysmd3
Entrez Gene ID
100767184
Gene Name
LysM, putative peptidoglycan-binding, domain containing 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614058: 388012 - 398771 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504391.2 XP_003504439.1 2 - 3 402 79
GenBank mRNA JP058582.1 - 1 - 2 250 79

View Full Product Details