Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc18
Entrez Gene ID
100771155
Gene Name
leucine rich repeat containing 18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613589: 2530615 - 2531382 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495313.1 XP_003495361.1 1 - 1 622 82

View Full Product Details