Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gdf9
Entrez Gene ID
100756618
Gene Name
growth differentiation factor 9
Gene Aliases
I79_004327
Chromosome Location
Chr.Un|NW_003613693: 293821 - 301370 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498484.2 XP_003498532.2 2 - 3 280 80
XM_007628466.1 XP_007626656.1 3 - 4 1276 80
XM_007628473.1 XP_007626663.1 3 - 4 1098 80
XM_007628479.1 XP_007626669.1 4 - 5 622 80

View Full Product Details