Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn6
Entrez Gene ID
100763564
Gene Name
lipocalin 6
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617557: 23091 - 27757 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515219.2 XP_003515267.2 2 - 3 234 107

View Full Product Details