Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chrm1
Entrez Gene ID
100755522
Gene Name
cholinergic receptor, muscarinic 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616404: 49305 - 50687 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654364.1 XP_007652554.1 1 - 1 1016 77

View Full Product Details