Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cckar
Entrez Gene ID
100774917
Gene Name
cholecystokinin A receptor
Gene Aliases
I79_021355
Chromosome Location
Chr.Un|NW_003615495: 82956 - 93865 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653033.1 XP_007651223.1 1 - 2 304 97

View Full Product Details