Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gsx1
Entrez Gene ID
100762769
Gene Name
GS homeobox 1
Gene Aliases
I79_002107
Chromosome Location
Chr.Un|NW_003613630: 2742141 - 2763816 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496674.1 XP_003496722.1 1 - 2 92 67

View Full Product Details