Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Etnk1
Entrez Gene ID
100765795
Gene Name
ethanolamine kinase 1
Gene Aliases
I79_004753
Chromosome Location
Chr.Un|NW_003613704: 2471714 - 2505221 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007633623.1 XP_007631813.1 1 - 2 54 73

View Full Product Details