Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magt1
Entrez Gene ID
100764570
Gene Name
magnesium transporter 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614139: 537120 - 576612 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505297.2 XP_003505345.1 1 - 2 160 63
GenBank mRNA JI885607.1 - 1 - 2 160 63
JP055298.1 - 1 - 2 160 63

View Full Product Details