Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc10a4
Entrez Gene ID
100753263
Gene Name
solute carrier family 10, member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615391: 138069 - 143399 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512203.1 XP_003512251.1 2 - 3 798 92

View Full Product Details