Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxd1
Entrez Gene ID
100757676
Gene Name
homeobox D1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613688: 1381904 - 1382625 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498334.1 XP_003498382.1 1 - 2 90 66

View Full Product Details