Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phlda2
Entrez Gene ID
100761127
Gene Name
pleckstrin homology-like domain, family A, member 2
Gene Aliases
I79_023756
Chromosome Location
Chr.Un|NW_003616688: 7013 - 8726 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654634.1 XP_007652824.1 1 - 2 355 76

View Full Product Details