Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pomp
Entrez Gene ID
100756796
Gene Name
proteasome maturation protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613630: 2046199 - 2059054 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496655.2 XP_003496703.1 4 - 5 373 69
GenBank mRNA JI875356.1 - 4 - 5 320 69
JP051129.1 - 4 - 5 318 69

View Full Product Details