Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxn2
Entrez Gene ID
100768700
Gene Name
forkhead box N2
Gene Aliases
I79_005646
Chromosome Location
Chr.Un|NW_003613744: 821301 - 861677 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499538.2 XP_003499586.1 2 - 3 719 107

View Full Product Details