Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g12b
Entrez Gene ID
100758369
Gene Name
phospholipase A2, group XIIB
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613999: 967210 - 980997 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503748.1 XP_003503796.1 1 - 2 211 74

View Full Product Details