Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lce6a
Entrez Gene ID
100755394
Gene Name
late cornified envelope 6A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614331: 30271 - 30516 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507098.1 XP_003507146.1 1 - 1 113 65

View Full Product Details