Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efcab4b
Entrez Gene ID
100763551
Gene Name
EF-hand calcium binding domain 4B
Gene Aliases
I79_001358
Chromosome Location
Chr.Un|NW_003613610: 1082122 - 1146591 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496083.2 XP_003496131.1 9 - 10 1152 106

View Full Product Details